Flask, Two-Neck, Pear-Shaped, Ace Glass Incorporated
Supplier: Ace Glass
9478-02 9478-10 9478-04
89074-906EA 115.13 CAD
89074-906 89074-910 89074-924
Flask, Two-Neck, Pear-Shaped, Ace Glass Incorporated
Flasks Round Bottom Flasks
These pear-shaped flasks have a side arm for use with a rubber stopper (not included)
The center joint is [ST] 14/20 outer. Fabricated from heavy wall borosilicate glass -- approximately 30% thicker than standard wall flasks.