Flask with Thermowell, Ace Glass Incorporated
Supplier: Ace Glass
6940-49 6940-51
89056-576EA 271.84 CAD
89056-576 89056-578
Flask with Thermowell, Ace Glass Incorporated
Flasks Round Bottom Flasks
This round-bottom flask has a center neck with a [SJ] 35/25 top joint and an angled side port thermometer well
Fabricated from borosilicate glass.