Dewar Flask, Plastic Coated, Ace Glass Incorporated
Supplier: Ace Glass
7076-15 7076-03 7076-12 7076-09 7076-06
89057-560EA 1524.53 CAD
89057-560 89057-562 89057-556 89057-554 89057-558
Dewar Flask, Plastic Coated, Ace Glass Incorporated
Dewars and Cryogenic Vessels
This cylindrical form, silvered, and evacuated Dewar is furnished with an aluminum base
Exterior, including the base, is coated with a tough, resilient, baked-on plastic which will not rupture if the flask breaks. Fabricated from borosilicate glass.