Reaction Flask with Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
6491-10 6491-20
89054-892EA 905.83 CAD
89054-892 89054-994
Reaction Flask with Stopcock, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This reaction flask has a 4mm bore glass stopcock on the outlet for rapid removal of contents without disturbing the general arrangement of the apparatus
The conical neck opening is 100mm (4-inches). Use a 89055-014 clamp to secure a matching 100mm head to the flask. Fabricated from borosilicate glass.
Ordering information: Clamp and head not included.