Flask, Three Necks, Zero Dead Space Valve, Ace Glass Incorporated
Supplier: Ace Glass
6483-110 6483-112 6483-120
89083-514EA 1083.56 CAD
89083-514 89083-516 89083-518
Flask, Three Necks, Zero Dead Space Valve, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This heavy wall, round-bottom flask features three [ST] 24/40 vertical necks and a 0-10 mm Zero Dead Space (ZDS™) valve as the bottom outlet
Fabricated from borosilicate glass.
Ordering information: Other [ST] neck joint sizes are available via special order. Contact your Avantor representative.
Order Now
SPECIFICATIONS
Environmentally Preferable