Reaction Flask, Spherical, with Indents, Ace Glass Incorporated
Supplier: Ace Glass
6481-05 6481-10 6481-15
89054-908EA 871.03 CAD
89054-908 89054-932 89054-934
Reaction Flask, Spherical, with Indents, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This rugged spherical flask has a conical neck opening of four inches (100mm)
It has side indents for improved stirring characteristics. Standard size flasks use regular Glas-Col heating mantles of equivalent capacity. The length of the straight section, including the flange, is approximately 9.5cm (3-3/4 inches). Not suitable for vacuum operations. Fabricated from borosilicate glass.
Ordering information: Clamp and gasket not included. Use a 89055-014 clamp to secure appropriate head to flask. For gasket, see 89055-004.