Gas Washing Bottle with Inner Coil, Ace Glass Incorporated
Supplier: Ace Glass
7167-30 7167-12 7167-07
89057-724EA 1599.73 CAD
89057-724 89057-726 89057-728
Gas Washing Bottle with Inner Coil, Ace Glass Incorporated
Bottles Gas Washing Bottles
This 270 ml gas washing bottle features an inner coil.
  • Fabricated from borosilicate glass
It is designed for use with a cyanide distillation apparatus.
Ordering information: Complete item includes bottle and inner coil. All components may also be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable