Flange, Duran®-Style, Ace Glass
Supplier: Ace Glass
15310-15 15310-20 15310-22 15310-25 15310-30 15311-16 15311-21 15311-23 15311-26 15311-31
89078-284EA 183.43 CAD
89078-284 89078-286 89078-288 89078-290 89078-292 89078-294 89078-296 89078-298 89078-300 89078-302
Flange, Duran®-Style, Ace Glass
Fittings and Connectors
This Duran®-style flange is used by glassblowers to fabricate reaction flasks.
It is available with or without an o-ring groove. For clamps, see 89055-114 (choose appropriate size). Fabricated from borosilicate glass.