Flask, with Septum Port, Ace Glass Incorporated
Supplier: Ace Glass
6933-224 6933-226 6933-227 6933-24 6933-26 6933-27 9461-10 9461-12 9461-14 9461-210 9461-212 9461-214
89056-498EA 163.24 CAD
89056-498 89056-500 89056-502 89056-504 89056-506 89056-508 89074-658 89074-660 89074-662 89074-664 89074-666 89074-668
Flask, with Septum Port, Ace Glass Incorporated
Flasks Round Bottom Flasks
This flask is used when handling air-sensitive materials
Available with standard walls or heavy walls (approximately 30% heavier than standard-wall flasks). With a [ST] outer joint on the center neck and a side septum joint for use with 89064-960 septum. Supplied with septum. [ST] 24/40 joints are reinforced. Fabricated from borosilicate glass.