Gas Washing Bottle with Large Disc, Ace Glass Incorporated
Supplier: Ace Glass
7162-02 7162-04 7162-06 7162-22 7162-24 7162-26 7162-50 7162-52 7162-54 7162-60 7162-62 7162-64 7162-12 7162-16 7162-70 7162-72 7162-74 7162-80 7162-82 7162-84 7162-14
89057-584EA 769.55 CAD
89057-584 89057-586 89057-588 89057-596 89057-598 89057-600 89057-602 89057-604 89057-606 89057-608 89057-610 89057-612 89057-632 89057-636 89057-686 89057-688 89057-690 89057-692 89057-694 89057-696 89057-714
Gas Washing Bottle with Large Disc, Ace Glass Incorporated
Bottles Gas Washing Bottles
The gas washing bottle with large disc provides greater capacity.
  • Fabricated from borosilicate glass
  • Joints are a [ST] 40/35
The 125 ml size has a 25 mm fritted disc, while the 250 and 500 ml sizes are fitted with a 30 mm disc. The inlet/outlet arms are 8 mm Ext.Ø.
Ordering information: Complete apparatus includes bottle and stopper. These items can also be purchased separately.