Streptomycin sulfate, powder

Supplier: Corning

61-088-RM 61-088-RM
CA45000-654EA 167.72 CAD
45000-654 CA45000-654
Streptomycin sulfate, powder
Streptomycin sulphate

Streptomycin is an antibiotic which acts as a protein synthesis inhibitor.


This aminoglycoside acts by binding to the 30S ribocomal subunit, preventing protein production and leading to bacterial cell death. Streptomycin is more effective against Gram-positive bacteria than Gram-negative and is offered in a powder form.

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
MDL Number: MFCD00037023
CAS Number: 3810-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR