Gooch Crucible, High Form, Ace Glass Incorporated
Supplier: Ace Glass
7170-02 7170-04 7170-06 7170-08 7170-10
89057-730EA 118.53 CAD
89057-730 89057-732 89057-734 89057-736 89057-738
Gooch Crucible, High Form, Ace Glass Incorporated
Crucibles
High Form Gooch crucible features an integral fritted disc that is 30mm in diameter
The height above the disc is 45mm. Available in five different porosities. Fabricated from borosilicate glass.