Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
21-040-CV 21-040-CM
CA45000-446PK 224.81 CAD
CA45000-446 CA45000-448
Corning® PBS (Phosphate-Buffered Saline), Corning
Buffers Cell Culture Buffers
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
PBS helps to maintain a constant pH and is isotonic and non-toxic to cells.