Allihn Tube with Frit, Ace Glass Incorporated
Supplier: Ace Glass
7194-02 7194-04 7194-06 7194-08
89058-270EA 150.51 CAD
89058-270 89058-272 89058-274 89058-276
Allihn Tube with Frit, Ace Glass Incorporated
Funnels Filtration Funnels Filter Funnels
The Allihn tube features an integral fritted disc with a diameter of 30mm
The height above the disc is 100mm, and the diameter of the bottom tube is 8mm. Available in four different porosities. Fabricated from borosilicate glass.