Penicillin : Streptomycin solution 100X, Corning®

Supplier: Corning
Danger

30-002-CI 30-002-CI
CA45000-652PK 262.59 CAD
CA45000-652 CA45000-653
Penicillin : Streptomycin solution 100X, Corning®
Penicillin/Streptomycin

This product is a mix of the antibiotics Penicillin (10,000 IU/mL) and Streptomycin (10,000 μg/ml) in a 100-fold working concentration. Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis. Penicillin-Streptomycin is effective against Gram-negative and Gram-positive bacteria.


Caution: Storage −25 to −15 °C. For research or further manufacturing use only. Not for use in diagnostic procedures.

CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)

Order Now

Specification Test Results

Formulation:
Penicillin G, potassium salt 10,000,000 U/L
Streptomycin sulfate 10.0 g/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR