IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
Supplier: Corning
Synonyms:
(2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol
, IPTG
, Isopropyl Beta-D-1-thiogalactopyranoside
46-102-RF
46-102-RF
CA45001-084EA
87.04
CAD
CA45001-084
45001-084
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
IPTG (Isopropyl-β-D-thiogalactopyranoside)
IPTG (isopropylthiogalactoside) is an inducer of the lac operon in bacteria, which is frequently used in cloning as a component of a recombinant plasmid.
When used with X-gal to detect lac gene activity, IPTG can be used for screening of clones containing a gene of interest in the lac operon.
Formula:
C₉H₁₈O₅S MW: 238.31 g/mol Boiling Pt: 402 °C (1013 hPa) Melting Pt: 109.5…110.5 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063273 CAS Number: 367-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...