Flask, Single Long Neck, Ace Glass Incorporated
Supplier: Ace Glass
6905-24 6905-26 6905-27
89056-420EA 116.31 CAD
89056-420 89056-422 89056-424
Flask, Single Long Neck, Ace Glass Incorporated
Flasks Round Bottom Flasks
This round-bottom flask has a single long neck with a reinforced [ST] 24/40 outer joint
Fabricated from borosilicate glass.