Boiling Flask Blanks, Quartz, Ace Glass Incorporated
Supplier: Ace Glass
6883-06 6883-08 6883-14 6883-16 6883-18 6883-20 6883-24
89056-156EA 503.03 CAD
89056-156 89056-158 89056-160 89056-162 89056-164 89056-166 89056-168
Boiling Flask Blanks, Quartz, Ace Glass Incorporated
Flasks Round Bottom Flasks
These round bottom flasks have a single unfinished center neck
These flask blanks are used by glassblowers in fabricating single- or multi-neck flasks. The neck O.D. listed is approximate. Fabricated from quartz.