G418 disulfate, powder, Corning®
Supplier: Corning
G418 Sulphate is recommended for use as a selection agent in a concentration range of 100 to 800 µg/ml. This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene. G418 is an aminoglycoside antibiotic similar in structure to gentamicin, neomycin and kanamycin. This antibiotic is effective against Gram negative and some Gram positive bacteria.
- Potency > 700 µg/mg
Caution: For Research or Further Manufacturing Use Only. Not for use in Diagnostic Procedures.
Formula:
C₂₀H₄₄N₄O₁₈S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00058314 CAS Number: 108321-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...