Ciprofloxacin hydrochloride, powder

Supplier: Corning

Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, 1-Cyclopropyl- 6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride

61-277-RF 61-277-RG
CA45000-620EA 292.39 CAD
CA45000-620 CA45000-622
Ciprofloxacin hydrochloride, powder
Ciprofloxacin hydrochloride

Ciprofloxacin Hydrochloride (HCl) is a 4-fluoroquinolone antibiotic often used to control mycoplasma contamination in infected cell lines


It works by interfering with the bacterial enzyme DNA gyrase, which is necessary for bacterial synthesis, replication, and transcription in both the active and non-active growth phases of the bacterial life cycle. Ciprofloxacin HCl is effective against a wide range of Gram-positive and Gram-negative bacteria and is offered in powder form.

Formula: C₁₇H₁₈FN₃O₃·HCl
MDL Number: MFCD00079044
CAS Number: 86393-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR