Low Form Zirconium Crucibles
Supplier: XRF Scientific
ZLF20 ZLF25 ZLF30 ZLF35 ZLF45 ZLF50 ZLF100
89401-368EA 239.59 CAD
89401-368 89401-370 89401-372 89401-374 89401-378 89401-380 89401-386
Low Form Zirconium Crucibles
Crucibles
Zirconium's distinctive properties make it a popular choice for fusions and fluxes in analytical chemistry and chemical processing.
These low form Zirconium crucibles are highly resistant to corrosion, and do not require handling with platinum tipped tongs. They can be cleaned with most common agents (except hydrofluoric acid), and suffer little to no attack or oxidation. Zirconium crucibles are resistant to melts of alkali carbonates, hydroxides, peroxides, borates, nitrate chlorides, and some fluorides. This construction virtually eliminates the problem of sample contamination. Lids areMore Product Information