Platinum Tipped Tongs, XRF Scientific
Supplier: XRF Scientific
PT99TAPL300
89401-338EA 2745.27 CAD
89401-338
Platinum Tipped Tongs, XRF Scientific
Tongs
Stainless steel tongs with 99.95% platinum tips are designed for use in manipulating platinum labware.
  • Tongs aid in eliminating contamination on platinum crucibles and dishes
  • Maintains the optimum conditions of platinum labware