Jackson Safety® G60 Level 3 Cut Resistant Gloves with Dyneema® Fiber, Kimberly-Clark
Supplier: Microflex
13823 13824 13825 13826 13827
CA89230-476CS 305.19 CAD
CA89230-476 CA89230-478 CA89230-480 CA89230-482 CA89230-484
Jackson Safety® G60 Level 3 Cut Resistant Gloves with Dyneema® Fiber, Kimberly-Clark
Gloves
Add cut resistance to your workers' gloves for tasks that require protection against hand lacerations with the new JACKSON SAFETY® G60 Level 3 Cut Resistant Gloves with Dyneema® fiber coated with polyurethane.
  • Dyneema® yarn with nylon, Lycra® fiber, and glass liner provides cut resistance and a comfortable fit
  • Excellent dexterity and dry and oil grip for improved productivity
  • Gray polyurethane coating enhances grip
  • Gray and salt and pepper color
  • Color coded hem for sizing
A great choice for working environments that pose a risk of minor cuts and abrasions. Typical uses include metal fabrication, glass handling, automotive assembly, stamping, and assembly.
Certifications: ANSI Level 3/EN 388 Level 3 cut resistance. Meets EN 420 including length.