SP Bel-Art NMR Sample Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18854-0005 F18854-0003
60986-335EA 151.33 CAD
60986-335 89217-376
SP Bel-Art NMR Sample Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Sturdy, stable polypropylene rack is made specifically for organizing and protecting thin-walled Nuclear Magnetic Resonance tubes.
  • Sturdy, stable polypropylene construction
  • Handles for easy transport
  • Chemical-resistant
  • Autoclavable
The lower deck has hemispherical wells to cradle tubes, and handles in the end plates make transport easy.
Racks stack securely to conserve bench space. Each rack holds up to 72 tubes in a 6x12 array. Chemical-resistant and autoclavable.