Ammonia solution 6.0 N, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Ammonia aqueousAmmonium hydroxide solutionAmmonia waterAmmonium hydroxide

BDHVBDH7641-2
BDH7641-2CS 7925.35 CAD
BDH7641-2
Ammonia solution 6.0 N, VWR Chemicals BDH®
Ammonia solution

41.4% (v/v) in water. Suitable as a titrant.

Formula: NH₃
MW: 35.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 5.980 - 6.020 N
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR