Potassium hydroxide solution 0.1 N in 2-propanol, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Caustic potash

BDHVBDH7630-1
BDH7630-1CS 3313.82 CAD
BDH7630-1
Potassium hydroxide solution 0.1 N in 2-propanol, VWR Chemicals BDH®
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear to turbid liquid
N.I.S.T. Traceable Yes
Normality 0.0995 - 0.1005 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR