Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
BDHVBDH7637-1
BDH7637-1CS
1787.83
CAD
BDH7637-1
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Hydrogen peroxide
Made with deionized water.
Formula:
H₂O₂ MW: 34.01 g/mol Boiling Pt: 100 °C (1013 hPa) Density: 1.01 g/cm³ (20 °C) |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. Traceable to SRM 8040 | Yes |
Assay | 3.00 - 3.50 % |
Stabilizer (Acetanilide) | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...