Potassium thiocyanate 0.1 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Potassium rhodanide

BDHVBDH7807-1
BDH7807-1CS 4101.7 CAD
BDH7807-1
Potassium thiocyanate 0.1 N in aqueous solution, VWR Chemicals BDH®
Potassium thiocyanate

Made with deionized water.

Formula: KSCN
MW: 97.18 g/mol
MDL Number: MFCD00011413
CAS Number: 333-20-0

Order Now

Specification Test Results

Normality 0.0995 - 0.1005 N
Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 919 Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR