Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
Caustic soda
BDH7994-4
BDH7994-6
BDH7994-4CS
593.81
CAD
BDH7994-4
BDH7994-6
BDH7994-7
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
Made with deionized water.
Formula:
NaOH MW: 40 g/mol Boiling Pt: 109 °C (1013 hPa) Density: ≤1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance/color | Clear liquid |
Shelf-life | 3 years |
Assay | 39.60 - 40.40 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...