Iodine 0.01 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

309V-06
BDH7897-1CS 1159.26 CAD
BDH7897-1
Iodine 0.01 N in aqueous solution, VWR Chemicals BDH®
Iodine

Made with iodine crystals, potassium iodide and deionized water.

Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3264
ADR: 8,II

Order Now

Specification Test Results

Normality 0.0095 - 0.0105 N
Appearance/color Clear dark brown liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR