Hydrochloric acid 20% (v/v), VWR Chemicals BDH®

Supplier: VWR International

BDH7871-4
BDH7871-4CS 709.13 CAD
BDH7871-4 CABDH7871-10
Hydrochloric acid 20% (v/v), VWR Chemicals BDH®
Hydrochloric acid

Per liter formula:
200mLs ACS HCL 36.5-38% W/W
800mLs Deionized Water

Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR