Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7690-1 BDH7690-3
BDH7690-1CS 2419.52 CAD
BDH7690-1 BDH7690-3
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Hydrogen peroxide
Formula: H₂O₂
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Ammonium (NH4) 5 ppm max.
Assay 29.0 - 32.0 %
Chloride (Cl) 3 ppm max.
Color (APHA) 10 max.
Heavy Metals (as Pb) 1 ppm max.
Iron (Fe) 0.5 ppm max.
Nitrate (NO3) 2 ppm max.
Phosphate (PO4) 2 ppm max.
Residue after evaporation 0.002 % max.
Sulfate (SO4) 5 ppm max.
Titratable Acid 0.0006 mEq/g
Stabilizer (sodium stannate) Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR