EDTA disodium salt 0.0100 M in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

BDH7621-1 BDH7621-4
BDH7621-1CS 976.76 CAD
BDH7621-1 BDH7621-4
EDTA disodium salt 0.0100 M in aqueous solution, VWR Chemicals BDH®
EDTA disodium salt

Suitable as a titrant. Made with deionized water.

Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 915 1
Molarity 0.0095 - 0.0105 M
Shelf-life 18 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR