Ammonia solution 5.0 N, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Ammonium hydroxide

BDHVBDH7615-1
BDH7615-1CS 1961.08 CAD
BDH7615-1
Ammonia solution 5.0 N, VWR Chemicals BDH®
Ammonia solution

Suitable as a titrant

Formula: NH₃
MW: 35.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 4.980 - 5.020 N
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR