Methyl orange 0,1% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

BDH7610-1 BDH7610-2
BDH7610-1CS 547 CAD
BDH7610-1 BDH7610-2
Methyl orange 0,1% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®
Methyl orange

Made with deionized water.

Formula: C₁₄H₁₄N₃NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3

Order Now

Specification Test Results

Appearance/color Clear orange liquid
Shelf-life 18 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR