Hydrochloric acid 0.2 N, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Hydrochloric acid solution, Muriatic acid

BDH7593-1 BDH7593-4
BDH7593-1CS 1148.09 CAD
BDH7593-1 BDH7593-4
Hydrochloric acid 0.2 N, VWR Chemicals BDH®
Hydrochloric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
MW: 36.46 g/mol
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 X
Normality 0.1995 - 0.2005 N
Shelf-life 18 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR