Ammonia solution 50% v/v zA, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
BDH7571-1
BDH7571-4
BDH7571-1CS
2548.87
CAD
BDH7571-1
BDH7571-4
Ammonia solution 50% v/v zA, VWR Chemicals BDH®
Ammonia solution
Per liter formula:
500mLs ACS Ammonium Hydroxide 28-30% W/W
500mLs Deionized Water
Formula:
NH₃ MW: 35.05 g/mol |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 |
Specification Test Results
Appearance/color | Clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...