Potassium hydroxide 0.1 N in methanol, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Potash caustic, Caustic potash

BDH7548-1 BDH7548-4
BDH7548-1CS 566.63 CAD
BDH7548-1 BDH7548-4
Potassium hydroxide 0.1 N in methanol, VWR Chemicals BDH®
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 X
Normality 0.0095 - 0.1005 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR