Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®

Supplier: VWR International

BDH7540-2 BDH7540-4
BDH7540-2CS 904.25 CAD
BDH7540-2 BDH7540-4
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 100 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable X
Shelf-life 18 months
Stabilizer (Acetanilide) Passes test
Assay 3.00 - 3.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR