EDTA disodium salt 0.05 M in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

BDH7497-1 BDH7497-4
BDH7497-1CS 1511.37 CAD
BDH7497-1 BDH7497-4
EDTA disodium salt 0.05 M in aqueous solution, VWR Chemicals BDH®
EDTA disodium salt

Made with deionized water.


Suitable as a titrant

Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 915 Yes
Molarity 0.0495 - 0.0505 M
Shelf-life 1 year


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR