Sodium hydroxide (5 - 60%) in aqueous solution 10% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Caustic soda

BDH7470-1 BDH7470-4
BDH7470-1CS 885.53 CAD
BDH7470-1 BDH7470-4
Sodium hydroxide (5 - 60%) in aqueous solution 10% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium hydroxide

Made with deionized water

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 2 years
Assay (w/v) 9.90 - 10.10 %
N.I.S.T. Traceable to SRM 84 Yes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR