Sodium azide 5% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR

Synonyms: Sodium triazide

BDHVBDH7465-2
BDH7465-2CS 1310.01 CAD
BDH7465-2
Sodium azide 5% (w/v) in aqueous solution, VWR Chemicals BDH®
Sodium azide

N.I.S.T. traceable solution, made with purified water.

Formula: NaN₃
MW: 65.01 g/mol
MDL Number: MFCD00003536
CAS Number: 26628-22-8
UN: 3287
ADR: 6.1,II

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 2 years
Assay (w/v) 4.90 - 5.10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR