Hydrochloric acid 5.0 N, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

BDH7419-1 BDH7419-4
BDH7419-1CS 847.28 CAD
BDH7419-1 BDH7419-4
Hydrochloric acid 5.0 N, VWR Chemicals BDH®
Hydrochloric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
MW: 36.46 g/mol
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
Normality 4.975 - 5.025 N
Shelf-life 2 years
Colouration (APHA) 10 max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR