Sodium hydroxide 33% (w/w) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Caustic soda

BDHVBDH7363-4
BDH7363-4CS 562.32 CAD
BDH7363-4
Sodium hydroxide 33% (w/w) in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

Sodium hydroxide 33% (w/w) in purified water.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
Assay 32.60 - 33.40 %
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR