Phenolphthalein 1% in alcohol, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
BDH7344-1
BDH7344-2
BDH7344-4
BDH7344-1CS
2420.22
CAD
BDH7344-1
BDH7344-2
BDH7344-4
Phenolphthalein 1% in alcohol, VWR Chemicals BDH®
Phenolphthalein
Made with denatured ethanol.
Formula:
C₂₀H₁₄O₄ MW: 318.33 g/mol Boiling Pt: ∼78 °C (1013 hPa) Density: ∼0.85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 UN: 1170 ADR: 3,II |
Specification Test Results
Appearance/color | Clear liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...