Phenolphthalein solution 0,5% analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
BDH7342-1
BDH7342-2
BDH7342-1CS
1006.48
CAD
BDH7342-1
BDH7342-2
Phenolphthalein solution 0,5% analytical reagent, VWR Chemicals BDH®
Phenolphthalein
Made with 50% isopropanol.
Formula:
C₂₀H₁₄O₄ MW: 318.33 g/mol |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 Merck Index: 14,07243 |
|
Specification Test Results
Appearance/color | Clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...