Fehling's reagent I (copper(II) sulfate solution) 3.46% (w/v), VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Cupric sulfate pentahydrateFehling A solutionCopper (II) sulfate 5-hydrate

BDHVBDH7316-1
BDH7316-1CS 1114.7 CAD
BDH7316-1
Fehling's reagent I (copper(II) sulfate solution) 3.46% (w/v), VWR Chemicals BDH®
Fehling's reagent I (Copper (II) sulphate solution)

Made with deionized water.

Formula: CuSO₄
MW: 159.61 g/mol
MDL Number: MFCD00010981
CAS Number: 7758-98-7
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Shelf-life 2 years
Appearance/color Clear blue liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR