Potassium hydroxide 45% (w/w) in aqueous solution, TECHNICAL, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7245-1 BDH7245-4
BDH7245-1CS 1878.58 CAD
BDH7245-1 BDH7245-4
Potassium hydroxide 45% (w/w) in aqueous solution, TECHNICAL, VWR Chemicals BDH®
Potassium hydroxide
Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Assay 43.00 - 47.00 %
Appearance Clear liquid
K₂CO₃ 0.2 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR