Potassium chloride saturated aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Potassiumchloride

BDH7242-1 BDH7242-2
BDH7242-1CS 1608.06 CAD
BDH7242-1 BDH7242-2
Potassium chloride saturated aqueous solution, VWR Chemicals BDH®
Potassium chloride

For refilling calomel reference electrodes. Ionic strength adjuster for calcium ion selective electrode.


  • Concentration is around 4 M

Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Appearance/color Clear liquid
Saturated product Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR