Hydrochloric acid 0.01 N, VWR Chemicals BDH®
Supplier: VWR International
BDH7237-1
BDH7237-1CS
948.86
CAD
BDH7237-1
Hydrochloric acid 0.01 N, VWR Chemicals BDH®
Hydrochloric acid
Made with high purity acid and deionized water. Suitable as a titrant.
Formula:
HCl Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,III |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. Traceable to SRM 723 | Yes |
Normality | 0.0095 - 0.0105 N |
Shelf-life | 1 year |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...